Thermometer, melting point, mercury filled, measuring range:-38:100/0.5C, enclosed scale, capillary lens front, colourless, dimens i on:250x6-7mm, immersion 30mm, Richter-top finish with button, suitable for government verification. Until 10th Octo 1 * 1 items
Thermometer, melting point, mercury filled, measuring range:-38:100/0.5C, enclosed scale, capillary lens front, colourless, dimens i on:250x6-7mm, immersion 30mm, Richter-top finish with button, suitable for government verification. Until 10th Octo 1 * 1 items