Charcoal activated
Glass bottle for solids, 500 g
Catalog # ROTH5963.3
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Carbon activated, Activated carbon, Activated charcoal
- CAS Number:7440-44-0
- Formula:C
- MDL Number:MFCD00133992
- Molecular Weight:12.01 g/mol
- Merck Index:13,01818
- Melting Pt:3550 °C
- Density:2 g/cm³ (20 °C)
- EINECS:231-153-3
- UN:1362
- ADR:4.2,III
- REACH:01-2119488894-16
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



