Closing cones, Combi-Stoppers
Closing cones for needle-free injection sites and seal pre-filled syringes, Blue
Supplier: B. Braun
Jump to:
Product Details & Documents
These PE Combi-Stoppers are used for sealing male or female Luer-Lock and Luer-Slip connections.
- Unique multifunctional design
- Sterile closing of pre-filled syringes and all kinds of open IV-accesses in IV-Sets
Available in red, blue and white.
Documents
Specifications
- Colour:Blue
- Description:Closing cones for needle-free injection sites and seal pre-filled syringes
- Seq:01