Order Entry
Export
Orders LinkContactUsLinkComponent
 

Human Granzyme B ELISPOT Set

Human Granzyme B ELISPOT set
Catalog # ABCAAB46581-480
Supplier:   Abcam
Human Granzyme B ELISPOT Set
Catalog # ABCAAB46581-480
Supplier:  Abcam
CAS Number:  
Human Granzyme B ELISPOT Set
Human Granzyme B ELISPOT Set

Human Granzyme B ELISPOT set

CAS Number:  
Price
1 110,00 €
Log in for availability
1 * 480 Tests
Product Details & Documents

Human Granzyme B ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Granzyme B production in a single cell suspension.

Specifications

Specifications

  • Description:
    Human Granzyme B ELISPOT set
  • Cat. No.:
    ABCAAB46581-480
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Species reactivity:
    Human
  • Storage temperature:
    +4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen name:
    Granzyme B

Specifications