Orders LinkContactUsLinkComponent
Automation tips for Tecan®, Axygen®
EV-200-R-S, 200 µl, +, -, Transparent, MCA96 / HV96, 50 racks, 96 each
  736-0450
 :   Corning
Automation tips for Tecan®, Axygen®
  736-0450
 :  Corning
Automation tips for Tecan®, Axygen®
Automation tips for Tecan®, Axygen®
EV-200-R-S, 200 µl, +, -, Transparent, MCA96 / HV96, 50 racks, 96 each
726,00 €
1 * 4.800 items
PP, with or without filter, sterile or non sterile.
  • Black tips are conductive for liquid level-sensing protocols
  • Optimised fine point for accurate dispensing
  • Certified RNase-, DNase- and endotoxin-free
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
 : Packaged in plastic blister packs.
 : * Economy packed in cardboard wrapper.
  • Packed:
    50 racks, 96 each
  • Sterile:
    +
  • Volume:
    200 µl
  • Seq:
    32
  • Tip style:
    EV-200-R-S
  • Tip colour:
    Transparent
  • For use with:
    MCA96 / HV96
  • Filter:
    -