Order Entry
Export
Orders LinkContactUsLinkComponent
4-Bromo-2,5-difluorophenol ≥99%

4-Bromo-2,5-difluorophenol ≥99%

Glass bottle for solids, 5 g
Catalog # H26212.06
Irritant
4-Bromo-2,5-difluorophenol ≥99%
Catalog # H26212.06
CAS Number:  486424-36-6
4-Bromo-2,5-difluorophenol ≥99%
4-Bromo-2,5-difluorophenol ≥99%

Glass bottle for solids, 5 g

CAS Number:  486424-36-6
Price
90,40 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 486424-36-6 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 Melting Point: 51-53° MDL No.: MFCD04973751

  • CAS Number:486424-36-6
  • Formula:C₆H₃BrF₂O
  • MDL Number:MFCD04973751
  • Molecular Weight:208.99 g/mol
  • Melting Pt:51…53 °C
  • Storage Temperature:Ambient
  • EINECS:None
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications