Zinc selenide
Glass bottle for solids, 50 g
CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Odour: Odourless Melting Point: 1100° Merck Reference: 14,10156 MDL No.: MFCD00011296
: Zinc (II) selenide
- :1315-09-9
- :SeZn
- :MFCD00011296
- :144.35 g/mol
- :14,10156
- :>1100 °C
- :5.42
- :Ambient
- :215-259-7
- :3283
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:50 g