Zinc selenide
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Odour: Odourless Melting Point: 1100° Merck Reference: 14,10156 MDL No.: MFCD00011296
Synonyms: Zinc (II) selenide
- CAS Number:1315-09-9
- Formula:SeZn
- MDL Number:MFCD00011296
- Molecular Weight:144.35 g/mol
- Merck Index:14,10156
- Melting Pt:>1100 °C
- Density:5.42
- Storage Temperature:Ambient
- EINECS:215-259-7
- UN:3283
- ADR:6.1,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



