Orders LinkContactUsLinkComponent
Lead(II) bromide ≥98%
Glass bottle for solids, 500 g
  A19406.36
Lead(II) bromide ≥98%
  A19406.36
 :  10031-22-8
Lead(II) bromide ≥98%
Lead(II) bromide ≥98%
Glass bottle for solids, 500 g
 :  10031-22-8
106,00 €
1 * 500 g
Purity: 98+% CAS No.: 10031-22-8 Molecular Formula: Br2Pb Formula Weight: 367.01 Odour: Odourless Melting Point: 373° Boiling Point: 916° Merck Reference: 14,5403 UN No.: 2291 MDL No.: MFCD00011156
 :  Lead dibromide, Lead(II) bromide
  •  :10031-22-8
  •  :Br₂Pb
  •  :MFCD00011156
  •  :367.01 g/mol
  •  :14,5403
  •  :916 °C
  •  :373 °C
  •  :6.66
  •  :Ambient
  •  :233-084-4
  •  :2291
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g