Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) bromide ≥98%

Lead(II) bromide ≥98%

Glass bottle for solids, 100 g
Catalog # A19406.22
Health
Irritant
Environment
Lead(II) bromide ≥98%
Catalog # A19406.22
CAS Number:  10031-22-8
Lead(II) bromide ≥98%
Lead(II) bromide ≥98%

Glass bottle for solids, 100 g

CAS Number:  10031-22-8
Price
35,40 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 10031-22-8 Molecular Formula: Br2Pb Formula Weight: 367.01 Odour: Odourless Melting Point: 373° Boiling Point: 916° Merck Reference: 14,5403 UN No.: 2291 MDL No.: MFCD00011156

Synonyms:  Lead dibromide, Lead(II) bromide
  • CAS Number:10031-22-8
  • Formula:Br₂Pb
  • MDL Number:MFCD00011156
  • Molecular Weight:367.01 g/mol
  • Merck Index:14,5403
  • Boiling Pt:916 °C
  • Melting Pt:373 °C
  • Density:6.66
  • Storage Temperature:Ambient
  • EINECS:233-084-4
  • UN:2291
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications