Order Entry
Export
Orders LinkContactUsLinkComponent
 

Soda lime indicator

Glass bottle for solids, 2.500 g
Catalog # A19352.0E
Corrosive
Soda lime indicator
Catalog # A19352.0E
CAS Number:  8006-28-8
Soda lime indicator
Soda lime indicator

Glass bottle for solids, 2.500 g

CAS Number:  8006-28-8
Price
153,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

CAS No.: 8006-28-8 Odour: Odourless Merck Reference: 14,8569 UN No.: 1907 MDL No.: MFCD00134094

  • CAS Number:8006-28-8
  • MDL Number:MFCD00134094
  • Merck Index:14,8569
  • Storage Temperature:Ambient
  • UN:1907
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications