Order Entry
Export
Orders LinkContactUsLinkComponent
 

3-Ethoxybenzaldehyde ≥97%

Glass bottle for solids, 2,5 g
Catalog # A18940.88
3-Ethoxybenzaldehyde ≥97%
Catalog # A18940.88
CAS Number:  22924-15-8
3-Ethoxybenzaldehyde ≥97%
3-Ethoxybenzaldehyde ≥97%

Glass bottle for solids, 2,5 g

CAS Number:  22924-15-8
Price
55,30 €
Log in for availability
1 * 2,5 g
Product Details & Documents

Purity: 97% CAS No.: 22924-15-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Boiling Point: 132-134/15mm MDL No.: MFCD00016606

  • CAS Number:22924-15-8
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00016606
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:132…134 °C (15 mmHg)
  • Melting Pt:21 °C
  • Density:1.07
  • Storage Temperature:Ambient
  • EINECS:245-333-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 g

Specifications