Orders LinkContactUsLinkComponent
Sulphamide ≥99%
Glass bottle for solids, 2,5 g
  A18680.88
Sulphamide ≥99%
  A18680.88
 :  7803-58-9
Sulphamide ≥99%
Sulphamide ≥99%
Glass bottle for solids, 2,5 g
 :  7803-58-9
44,80 €
1 * 2,5 g
Purity: 99% CAS No.: 7803-58-9 Molecular Formula: SO2(NH2)2 Formula Weight: 96.11 Melting Point: 91-92° Solubility: Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol MDL No.: MFCD00011606
 :  Sulphuric diamide
  •  :7803-58-9
  •  :H₄N₂O₂S
  •  :MFCD00011606
  •  :96.11 g/mol
  •  :14,8922
  •  :91…92 °C
  •  :1.61
  •  :Ambient
  •  :232-262-9
  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 g