Order Entry
Export
Orders LinkContactUsLinkComponent
Sulphamide ≥99%

Sulphamide ≥99%

Glass bottle for solids, 50 g
Catalog # A18680.18
Irritant
Sulphamide ≥99%
Catalog # A18680.18
CAS Number:  7803-58-9
Sulphamide ≥99%
Sulphamide ≥99%

Glass bottle for solids, 50 g

CAS Number:  7803-58-9
Price
356,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 99% CAS No.: 7803-58-9 Molecular Formula: SO2(NH2)2 Formula Weight: 96.11 Melting Point: 91-92° Solubility: Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol MDL No.: MFCD00011606

Synonyms:  Sulphuric diamide
  • CAS Number:7803-58-9
  • Formula:H₄N₂O₂S
  • MDL Number:MFCD00011606
  • Molecular Weight:96.11 g/mol
  • Merck Index:14,8922
  • Melting Pt:91…92 °C
  • Density:1.61
  • Storage Temperature:Ambient
  • EINECS:232-262-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications