Sulphamide ≥99%
Glass bottle for solids, 10 g
Catalog # A18680.09
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 7803-58-9 Molecular Formula: SO2(NH2)2 Formula Weight: 96.11 Melting Point: 91-92° Solubility: Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol MDL No.: MFCD00011606
Synonyms: Sulphuric diamide
- CAS Number:7803-58-9
- Formula:H₄N₂O₂S
- MDL Number:MFCD00011606
- Molecular Weight:96.11 g/mol
- Merck Index:14,8922
- Melting Pt:91…92 °C
- Density:1.61
- Storage Temperature:Ambient
- EINECS:232-262-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



