Sodium hydroxide 98%, soluble
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Sodium hydroxide 98%, soluble
Catalog # A18395.36
Supplier: Thermo Fisher Scientific
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odour: Odourless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,8627
- Boiling Pt:1390 °C
- Melting Pt:318 °C
- Density:2.13
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



