Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium hydroxide 98%, soluble

Sodium hydroxide 98%, soluble

Glass bottle for solids, 2.500 g
Catalog # A18395.0E
Corrosive
Sodium hydroxide 98%, soluble
Catalog # A18395.0E
CAS Number:  1310-73-2
Sodium hydroxide 98%, soluble
Sodium hydroxide 98%, soluble

Glass bottle for solids, 2.500 g

CAS Number:  1310-73-2
Price
107,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 98% CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Odour: Odourless pH: 14 Melting Point: 318° Boiling Point: 1390° Merck Reference: 14,8627 UN No.: 1823 MDL No.: MFCD00003548

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,8627
  • Boiling Pt:1390 °C
  • Melting Pt:318 °C
  • Density:2.13
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications