Cellulose, microcrystalline
Glass bottle for solids, 2.500 g
Supplier: Thermo Fisher Scientific
Cellulose, microcrystalline
Catalog # A17730.0E
Supplier: Thermo Fisher Scientific
CAS Number: 9004-34-6
Jump to:
Product Details & Documents
CAS No.: 9004-34-6 Physical Form: Powder Appearance: White Merck Reference: 14,1965 MDL No.: MFCD00081512
Synonyms: Microcrystallinecellulose
- CAS Number:9004-34-6
- Formula:(C₁₂H₂₂O₁₁)n
- MDL Number:MFCD00081512
- Molecular Weight:342.3 g/mol
- Merck Index:14,1965
- Melting Pt:260…270 °C
- Density:1.27…1.6 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-674-9
Documents
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Cell Culture Workflow
Product Literature
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2.500 g



