Order Entry
Export
Orders LinkContactUsLinkComponent
Cellulose, microcrystalline

Cellulose, microcrystalline

Glass bottle for solids, 10.000 g
Catalog # A17730.0C
Irritant
Cellulose, microcrystalline
Catalog # A17730.0C
CAS Number:  9004-34-6
Cellulose, microcrystalline
Cellulose, microcrystalline

Glass bottle for solids, 10.000 g

CAS Number:  9004-34-6
Price
1 050,00 €
Log in for availability
1 * 10.000 g
Product Details & Documents

CAS No.: 9004-34-6 Physical Form: Powder Appearance: White Merck Reference: 14,1965 MDL No.: MFCD00081512

Synonyms:  Microcrystallinecellulose
  • CAS Number:9004-34-6
  • Formula:(C₁₂H₂₂O₁₁)n
  • MDL Number:MFCD00081512
  • Molecular Weight:342.3 g/mol
  • Merck Index:14,1965
  • Melting Pt:260…270 °C
  • Density:1.27…1.6 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-674-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10.000 g

Specifications