Order Entry
Export
Orders LinkContactUsLinkComponent
Oleic acid tech. 90%, Technical Grade

Oleic acid tech. 90%, Technical Grade

Glass bottle, 1.000 mL
Catalog # A16663.AU
Irritant
Oleic acid tech. 90%, Technical Grade
Catalog # A16663.AU
CAS Number:  112-80-1
Oleic acid tech. 90%, Technical Grade
Oleic acid tech. 90%, Technical Grade

Glass bottle, 1.000 mL

CAS Number:  112-80-1
Price
52,90 €
Log in for availability
1 * 1.000 mL
Product Details & Documents

Purity: 90% CAS No.: 112-80-1 Molecular Formula: C18H34O2 Formula Weight: 282.47 Storage Temperature: 2°C to 8°C Physical Form: Clear liquid Density: 0.887 Refractive Index: 1.4610 Sensitivity: Air Sensitive Appearance: Colourless Melting Point: 13-14° Boiling Point: 286°/100mm Beilstein Reference: 1726542 Merck Reference: 14,6828 MDL No.: MFCD00064242

Synonyms:  (9Z)-9-Octadecenoic acid
  • CAS Number:112-80-1
  • Formula:C₁₈H₃₄O₂
  • MDL Number:MFCD00064242
  • Molecular Weight:282.47 g/mol
  • Merck Index:14,6828
  • Boiling Pt:286 °C (100 mmHg)
  • Melting Pt:13…14 °C
  • Density:0.887
  • Flash Pt:180 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-007-1
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1.000 mL

Specifications