Solvent Blue 38
100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.
CAS No.: 1328-51-4 Odour: Odourless Melting Point: ca 235° dec. MDL No.: MFCD00071424
Synonyms: Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN
- CAS Number:1328-51-4
- Formula:C32H12CuN8Na2O6S2
- MDL Number:MFCD00071424
- Molecular Weight:778.15 g/mol
- Melting Pt:∼ 235 °C (dec.)
- Storage Temperature:Ambient
- EINECS:215-523-1
Specifications
Specifications
- Pk:100 g



