Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dibromopropane ≥98%

1,3-Dibromopropane ≥98%

Glass bottle for solids, 5.000 g
Catalog # A14687.0I
Flammable
Irritant
Environment
1,3-Dibromopropane ≥98%
Catalog # A14687.0I
CAS Number:  109-64-8
1,3-Dibromopropane ≥98%
1,3-Dibromopropane ≥98%

Glass bottle for solids, 5.000 g

CAS Number:  109-64-8
Price
743,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 98% CAS No.: 109-64-8 Molecular Formula: C3H6Br2 Formula Weight: 201.90 Flash Point: 54°(130°F) Physical Form: Liquid Density: 1.99 Refractive Index: 1.5230 Appearance: Colourless Melting Point: -34° Boiling Point: 167° Beilstein Reference: 635662 Merck Reference: 14,9713 UN No.: 1993 MDL No.: MFCD00000255

  • CAS Number:109-64-8
  • Formula:BrH₂CCH₂CH₂Br
  • MDL Number:MFCD00000255
  • Molecular Weight:201.89 g/mol
  • Merck Index:14,9713
  • Boiling Pt:167 °C
  • Melting Pt:–34 °C
  • Density:1.99
  • Flash Pt:54 °C (130 °F)
  • Storage Temperature:Ambient
  • EINECS:203-690-3
  • UN:1993
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications