Order Entry
Export
Orders LinkContactUsLinkComponent
 

Brilliant green

Glass bottle for solids, 100 g
Catalog # A12801.22
Irritant
Brilliant green
Catalog # A12801.22
CAS Number:  633-03-4
Brilliant green
Brilliant green

Glass bottle for solids, 100 g

CAS Number:  633-03-4
Price
54,80 €
Log in for availability
1 * 100 g
Product Details & Documents

CAS No.: 633-03-4 Molecular Formula: C27H34N2O4S Formula Weight: 482.65 Melting Point: 210° dec. Beilstein Reference: 3901207 Merck Reference: 14,1374 MDL No.: MFCD00011880

Synonyms:  Ethyl Green, CI 42040, Basic Green 1, N-(4-((4-(Diethylamino)phenyl)(phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium hydrogen sulphate, Diamond Green, Malachite Green G, Emerald Green, Astradiamant green GX, Solid Green JO, Brilliant Green hydrogen sulphate
  • CAS Number:633-03-4
  • Formula:C₂₇H₃₄N₂O₄S
  • MDL Number:MFCD00011880
  • Molecular Weight:482.65 g/mol
  • Merck Index:14,1374
  • Melting Pt:210 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:211-190-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications