Cyclohexanone-2,4-dinitrophenylhydrazone 99%
5 g
Supplier: Thermo Fisher Scientific
Cyclohexanone-2,4-dinitrophenylhydrazone 99%
Catalog # A12203.06
Supplier: Thermo Fisher Scientific
CAS Number: 1589-62-4
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 1589-62-4 Molecular Formula: C12H14N4O4 Formula Weight: 278.27 Melting Point: 158-160° MDL No.: MFCD00001658
- CAS Number:1589-62-4
- Formula:C₁₂H₁₄N₄O₄
- MDL Number:MFCD00001658
- Molecular Weight:278.27 g/mol
- Melting Pt:158…160 °C
- Storage Temperature:Ambient
- EINECS:216-458-1
Specifications
Specifications
- Pk:5 g



