Order Entry
Export
Orders LinkContactUsLinkComponent
Cyclohexanone-2,4-dinitrophenylhydrazone 99%

Cyclohexanone-2,4-dinitrophenylhydrazone 99%

1 g
Catalog # A12203.03
Cyclohexanone-2,4-dinitrophenylhydrazone 99%
Catalog # A12203.03
CAS Number:  1589-62-4
Cyclohexanone-2,4-dinitrophenylhydrazone 99%
Cyclohexanone-2,4-dinitrophenylhydrazone 99%

1 g

CAS Number:  1589-62-4
Price
60,40 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 99% CAS No.: 1589-62-4 Molecular Formula: C12H14N4O4 Formula Weight: 278.27 Melting Point: 158-160° MDL No.: MFCD00001658

  • CAS Number:1589-62-4
  • Formula:C₁₂H₁₄N₄O₄
  • MDL Number:MFCD00001658
  • Molecular Weight:278.27 g/mol
  • Melting Pt:158…160 °C
  • Storage Temperature:Ambient
  • EINECS:216-458-1
Specifications

Specifications

  • Pk:
    1 g

Specifications