Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) acetate trihydrate ≥99%

Lead(II) acetate trihydrate ≥99%

Glass bottle for solids, 250 g
Catalog # A11746.30
Health
Environment
Lead(II) acetate trihydrate ≥99%
Catalog # A11746.30
CAS Number:  6080-56-4
Lead(II) acetate trihydrate ≥99%
Lead(II) acetate trihydrate ≥99%

Glass bottle for solids, 250 g

CAS Number:  6080-56-4
Price
31,10 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 6080-56-4 Molecular Formula: Pb(OOCCH3)2.3H2O Formula Weight: 379.33 (325.28anhy) Melting Point: 75° Beilstein Reference: 3730298 Merck Reference: 14,5397 UN No.: 1616 MDL No.: MFCD00150023

Synonyms:  Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
  • CAS Number:6080-56-4
  • Formula:(H₃CCOO)₂Pb·3H₂O
  • MDL Number:MFCD00150023
  • Molecular Weight:379.34 g/mol
  • Merck Index:14,5397
  • Boiling Pt:280 °C (1013 hPa)
  • Melting Pt:75 °C
  • Density:2.55
  • Storage Temperature:Ambient
  • EINECS:206-104-4
  • UN:1616
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications