Orders LinkContactUsLinkComponent
Lead(II) acetate trihydrate ≥99%
Glass bottle for solids, 1.000 g
  A11746.0B
Lead(II) acetate trihydrate ≥99%
  A11746.0B
 :  6080-56-4
Lead(II) acetate trihydrate ≥99%
Lead(II) acetate trihydrate ≥99%
Glass bottle for solids, 1.000 g
 :  6080-56-4
90,80 €
1 * 1.000 g
Purity: 99% CAS No.: 6080-56-4 Molecular Formula: Pb(OOCCH3)2.3H2O Formula Weight: 379.33 (325.28anhy) Melting Point: 75° Beilstein Reference: 3730298 Merck Reference: 14,5397 UN No.: 1616 MDL No.: MFCD00150023
 :  Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
  •  :6080-56-4
  •  :(H₃CCOO)₂Pb·3H₂O
  •  :MFCD00150023
  •  :379.34 g/mol
  •  :14,5397
  •  :280 °C (1013 hPa)
  •  :75 °C
  •  :2.55
  •  :Ambient
  •  :206-104-4
  •  :1616
  •  :6.1,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g