Order Entry
Export
Orders LinkContactUsLinkComponent
2-Octanone ≥98%

2-Octanone ≥98%

Glass bottle, 5.000 mL
Catalog # A11643.0J
Flammable
Irritant
2-Octanone ≥98%
Catalog # A11643.0J
CAS Number:  111-13-7
2-Octanone ≥98%
2-Octanone ≥98%

Glass bottle, 5.000 mL

CAS Number:  111-13-7
Price
278,00 €
Log in for availability
1 * 5.000 mL
Product Details & Documents

Purity: 98% CAS No.: 111-13-7 Molecular Formula: CH3(CH2)5C(O)CH3 Formula Weight: 128.22 Flash Point: 63°(145°F) Refractive Index: 1.4150 Melting Point: -16° Boiling Point: 172-173° Beilstein Reference: 635843 Merck Reference: 14,4711 MDL No.: MFCD00009540

Synonyms:  Octan-2-one
  • CAS Number:111-13-7
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00009540
  • Molecular Weight:128.21 g/mol
  • Merck Index:14,4711
  • Boiling Pt:172…173 °C
  • Melting Pt:–16 °C
  • Density:0.819
  • Flash Pt:54 °C (129 °F)
  • Storage Temperature:Ambient
  • EINECS:203-837-1
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5.000 mL

Specifications