2-Octanone ≥98%
Glass bottle, 1.000 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 111-13-7 Molecular Formula: CH3(CH2)5C(O)CH3 Formula Weight: 128.22 Flash Point: 63°(145°F) Refractive Index: 1.4150 Melting Point: -16° Boiling Point: 172-173° Beilstein Reference: 635843 Merck Reference: 14,4711 MDL No.: MFCD00009540
Synonyms: Octan-2-one
- CAS Number:111-13-7
- Formula:C₈H₁₆O
- MDL Number:MFCD00009540
- Molecular Weight:128.21 g/mol
- Merck Index:14,4711
- Boiling Pt:172…173 °C
- Melting Pt:–16 °C
- Density:0.819
- Flash Pt:54 °C (129 °F)
- Storage Temperature:Ambient
- EINECS:203-837-1
- UN:1224
- ADR:3,III
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1.000 mL



