Counting chambers, Malassez
Counting chamber with double ruling, 0,20 mm
Catalog # HECH40453702
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
These counting chambers are designed for blood tests and urine analysis.
Specifications
- Depth of chamber:0,20 mm
- Description:Counting chamber with double ruling