Folic acid
Glass bottle for solids, 25 g
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9
- CAS Number:59-30-3
- Formula:C₁₉H₁₉N₇O₆
- MDL Number:MFCD00079305
- Molecular Weight:441.41 g/mol
- EINECS:200-419-0
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



