Universal couplings, BOLA
internal thread M 6, female, UNF ¼" 28 G, outer thread, PTFE
Catalog # BOHLF770-08
Supplier: Bohlender
Jump to:
Product Details & Documents
PTFE, white.
- Temperature resistance: from –200 to +250 °C
- Universal chemical resistance
- FDA conform
Coupling for transition from M6 thread to UNF ¼" 28 G thread. The M6 thread is marked by a groove.
Specifications
- Description:internal thread M 6
- Material:PTFE
- Thread:female, UNF ¼" 28 G, outer thread