Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer), Sigma-Aldrich®
TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), 4 L
Catalog # SIALT9650-4L
Supplier: Merck
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer), Sigma-Aldrich®
Catalog # SIALT9650-4L
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:4 L
- Description:TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)
- Cat. No.:SIALT9650-4L
- M-Clarity quality level:MQ200



