Order Entry
Export
Orders LinkContactUsLinkComponent
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®

Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L
Catalog # SIALT4415-1L
Supplier:   Merck
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®
Catalog # SIALT4415-1L
Supplier:  Merck
CAS Number:  
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer), Sigma-Aldrich®

TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), Glass bottle, 1 L

CAS Number:  
Price
82,40 €
Log in for availability
1 * 1 L
Product Details & Documents

TBE (Tris/Borate/EDTA) Buffer is commonly used in nucleic acid electrophoresis. This solution is effective under slightly basic conditions, which keeps DNA deprotonated, water-soluble, and protected from degradation. This concentrate can be easily diluted to 1X or 0,5X before use (with molecular biology grade water).

TBE running buffer is the most commonly used buffer for DNA and RNA polyacrylamide gel electrophoresis. TBE is used with non-denaturing or denaturing (7 M urea) gels. It is also routinely used for DNA automated sequencing gel. TBE can also be used for agarose gels but is not recommended for preparative gels for recovery of nucleic acids.

Dilution of the TBE stock concentrates to a 1X TBE running buffer results in a buffer containing 89 mM Tris-borate and 2 mM EDTA, pH 8,3. The 5X or 10X stocks may also be added to an acrylamide/bis-acrylamide stock solution for making the PAGE gel. Applied voltages of less than 5 V/cm (distance between the electrodes of the unit) are recommended for maximum resolution.

Packaging: The 4 litre, 10 litre and 20 litre sizes are supplied in dispenser with a spigot.

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)
  • Cat. No.:
    SIALT4415-1L
  • M-Clarity quality level:
    MQ200

Specifications