D-(-)-Sorbitol, Sigma-Aldrich®
Glass bottle for solids, 5 kg
SIALS6021-5KG
: Merck
: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- :50-70-4
- :HOH₂C(CH(OH))₄CH₂OH
- :MFCD00004708
- :182.17 g/mol
- :13,08797
- :295 °C (1013 hPa)
- :94…98 °C
- :1.524 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- :>100 °C
- :Ambient
- :200-061-5
- Pack type:Glass bottle for solids
- Pk:5 kg