Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ethidium bromide, Sigma-Aldrich®

Glass bottle for solids, 25 g
Catalog # SIALE8751-25G
Toxic
Health
Supplier:   Merck
Ethidium bromide, Sigma-Aldrich®
Catalog # SIALE8751-25G
Supplier:  Merck
CAS Number:  1239-45-8
Ethidium bromide, Sigma-Aldrich®

Glass bottle for solids, 25 g

CAS Number:  1239-45-8
Price
826,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0.1 mmHg)
  • Melting Pt:261…264 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:<100 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,I
  •  :M-Clarity™ quality level = MQ200
  • EINECS:214-984-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications