Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,4-Dithioerythritol, Sigma-Aldrich®

Glass bottle for solids, 10 g
Catalog # SIALD8161-10G
Irritant
Supplier:   Merck
1,4-Dithioerythritol, Sigma-Aldrich®
Catalog # SIALD8161-10G
Supplier:  Merck
CAS Number:  6892-68-8
1,4-Dithioerythritol, Sigma-Aldrich®

Glass bottle for solids, 10 g

CAS Number:  6892-68-8
Price
428,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
  • CAS Number:6892-68-8
  • Formula:C₄H₁₀O₂S₂
  • MDL Number:MFCD00063750
  • Molecular Weight:154.25 g/mol
  • Boiling Pt:286 °C (1013 hPa)
  • Melting Pt:82…83 °C
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:229-998-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications