Orders LinkContactUsLinkComponent
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Makro, 1000 - 5000 µl, -, Transparent, Gilson® Pipetman®/Neo, 50/rack
Catalog # RATI2410630
Supplier:   RATIOLAB
Pipette tips, ratiolab®
Catalog # RATI2410630
Supplier:  RATIOLAB
Pipette tips, ratiolab®
Pipette tips, ratiolab®
Makro, 1000 - 5000 µl, -, Transparent, Gilson® Pipetman®/Neo, 50/rack
Price
26,00 €
Log in for availability
1 * 50 items
Product Details & Documents
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
  • Made of high quality PP
  • Tight fit due to the precisely fitting cone
  • Guaranteed dust-free due to special ionization treatment
  • Excellent transparency due to high quality surfaces
  • Minimal residual wetting due to controlled quality of raw materials
  • Excellent volume accuracy due to precise tip orifice
  • MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
Specifications
  • Packed:
    50/rack
  • Sterile:
    -
  • Volume:
    1000 - 5000 µl
  • Seq:
    37
  • Tip style:
    Makro
  • Tip colour:
    Transparent
  • For use with:
    Gilson® Pipetman®/Neo