Order Entry
Export
Orders LinkContactUsLinkComponent
 

Thermal transfer printer labels, type B-8423

Printer labels, B-8423, BPT-6-8423-2.5, Polyester, Permanent acrylic, Thermal transfer, White, BBP12, 38×19 mm, 2500 labels/roll
Catalog # BRDY236715
Supplier:   Brady
Thermal transfer printer labels, type B-8423
Catalog # BRDY236715
Supplier:  Brady
Thermal transfer printer labels, type B-8423
Thermal transfer printer labels, type B-8423

Printer labels, B-8423, BPT-6-8423-2.5, Polyester, Permanent acrylic, Thermal transfer, White, BBP12, 38×19 mm, 2500 labels/roll

Price
128,00 €
Log in for availability
1 * 1 Roll
Product Details & Documents

Printable satin polyester labels for thermal transfer printers.

  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves

This material is UL and CSA approved with its respective ribbon. See the recommended ribbon series for the appropriate ribbon to use.

Specifications
  • Printer model:
    BBP12
  • Printing method:
    Thermal transfer
  • Type:
    B-8423
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    38×19 mm
  • Material:
    Polyester
  • Seq:
    07
  • Model:
    BPT-6-8423-2.5
  • Packed:
    2500 labels/roll
  • Adhesive:
    Permanent acrylic