Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
200 mm
Catalog # HECH49050021
Funnels with short stem
Catalog # HECH49050021
Funnels with short stem
Funnels with short stem
200 mm
Price
8,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
  • Good chemical resistance
  • Suspension eye for easy storage
  • Easy to clean
Specifications
  • Funnel O-Ø:
    200 mm