Funnels with short stem
120 mm
Catalog # HECH49050012
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
- Good chemical resistance
- Suspension eye for easy storage
- Easy to clean
Specifications
- Funnel O-Ø:120 mm