Orders LinkContactUsLinkComponent
Flasks, Erlenmeyer, with standard ground joint
50 ml, 29/32 NS
  HECH41918012
Flasks, Erlenmeyer, with standard ground joint
  HECH41918012
Flasks, Erlenmeyer, with standard ground joint
Flasks, Erlenmeyer, with standard ground joint
50 ml, 29/32 NS
19,90 €
1 * 1 items
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.
  • Socket size:
    29/32 NS
  • Capacity:
    50 ml