Orders LinkContactUsLinkComponent
Flasks, Erlenmeyer, with standard ground joint
250 ml, 45/40 NS
  HECH41915034
Flasks, Erlenmeyer, with standard ground joint
  HECH41915034
Flasks, Erlenmeyer, with standard ground joint
Flasks, Erlenmeyer, with standard ground joint
250 ml, 45/40 NS
31,60 €
1 * 1 items
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.
  • Socket size:
    45/40 NS
  • Capacity:
    250 ml