Volumetric flasks, class A, blue markings
100 ml, 12/21, 0,10 ± ml
Jump to:
Borosilicate glass 3.3.
- Calibrated to contain (TC, In)
- Marks and inscriptions in blue
- Conformity certified (DE-M marking)
: DIN EN ISO 1042
Documents
Certificates
Search by Lot NumberSpecifications
- NS:12/21
- Capacity:100 ml
- Seq:07
- Tolerance:0,10 ± ml