Orders LinkContactUsLinkComponent
Cover glasses, square
Cover glasses, square
22×22 mm, #1, Borosilicate glass
Catalog # HECH41000115
Cover glasses, square
Catalog # HECH41000115
Cover glasses, square
Cover glasses, square
22×22 mm, #1, Borosilicate glass
Price
32,90 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Borosilicate glass coverslips for histology and microscopy.
Specifications
  • No.:
    #1
  • L×W:
    22×22 mm
  • Material:
    Borosilicate glass