Clips for conical ground joints
Orange, 34/35
Catalog # HECH42386034
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
Specifications
- For NS:34/35
- Colour:Orange
- Seq:07