Clips for conical ground joints
Purple, 12/21
Catalog # HECH42386012
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
Specifications
- For NS:12/21
- Colour:Purple
- Seq:02