Clips for conical ground joints
Green, 10/19
Catalog # HECH42386010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
Specifications
- For NS:10/19
- Colour:Green
- Seq:01