Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
250 ml, 29/32 NS
Catalog # HECH41911018
Flasks, flat bottom with standard ground joint
Catalog # HECH41911018
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
250 ml, 29/32 NS
Price
23,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Specifications
  • Socket size:
    29/32 NS
  • Capacity:
    250 ml
  • Seq:
    04