Order Entry
Export
Orders LinkContactUsLinkComponent
SPE cartridge, Sep-Pak®

SPE cartridge, Sep-Pak®

Plus short cartridge, Cartridge, DNPH-Silica, 125 Å, 55 - 105 µm, 0,7 ml, 350 mg
Catalog # WATEWAT037500
Supplier:   WATERS
SPE cartridge, Sep-Pak®
Catalog # WATEWAT037500
Supplier:  WATERS
SPE cartridge, Sep-Pak®

Plus short cartridge, Cartridge, DNPH-Silica, 125 Å, 55 - 105 µm, 0,7 ml, 350 mg

Price
615,00 €
Log in for availability
1 * 20 items
Product Details & Documents

Specially designed for analysing formaldehyde, other aldehydes and ketones in air.

Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    55 - 105 µm
  • Pore size:
    125 Å
  • Capacity:
    0,7 ml
  • Description:
    Plus short cartridge
  • Bed weight:
    350 mg
  • Phase:
    DNPH-Silica